Imprimer
Magnetic stirring bars, PTFE, with crosshead on both sides, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C360-04 C360-25 C360-22 C360-19 C360-10 C360-16 C360-07 C360-13
BOHLC360-04EA 6 EUR
BOHLC360-04 BOHLC360-25 BOHLC360-22 BOHLC360-19 BOHLC360-10 BOHLC360-16 BOHLC360-07 BOHLC360-13
Magnetic stirring bars, PTFE, with crosshead on both sides, BOLA
Agitateurs Barreaux magnétiques
Magnetic stirrer bars with magnetic core of Alnico 5 encapsulated in PTFE. Cross heads are present on both sides.
  • Chemical resistance
  • Symmetrical fins most preferable for tall, narrow diameter vessels
Temperature resistance: −200 to +250 °C.