Imprimer
Evaporating dishes, cylindrical, transparent, without spout, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A177-03 A177-01
BOHLA177-03EA 203 EUR
BOHLA177-03 BOHLA177-01
Evaporating dishes, cylindrical, transparent, without spout, BOLA
Plateaux/Récipients/Pots Capsules d'évaporation
Evaporating dishes, PFA, with spout.
  • Transparent, non-porous, stackable
  • Universal chemical resistance
  • Temperarture resistance from −200 to 250 °C