Imprimer
Evaporating dishes, cylindrical, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A170-01 A170-02 A170-03 A170-04 A176-03 A176-02
BOHLA170-01EA 38.6 EUR
BOHLA170-01 BOHLA170-02 BOHLA170-03 BOHLA170-04 BOHLA176-03 BOHLA176-02
Evaporating dishes, cylindrical, BOLA
Plateaux/Récipients/Pots Capsules d'évaporation
Evaporating dishes are made of PTFE.
  • Temperature resistance from −200 to 250 °C
  • With universal chemical resistance