Imprimer
Centrifuge tubes, Axygen®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SCT-15ML-500 SCT-50ML-500 SCT-15ML-R-S SCT-15ML-25-S SCT-50ML-25-S SCT-50ML-R-S
525-0518EA 152 EUR
525-0518 525-0519 525-1746 525-0780 525-0781 525-1747
Centrifuge tubes, Axygen®
Tubes Tubes à centrifuger
PP, conical, printed graduation, with HDPE flat screw caps.
  • Max. RCF: 12 000×g
  • Easy one hand operation
  • Large writing area for sample identification