Chlorure de tris(triphénylphosphine)ruthenium (II) ≥99.95% (base métaux) Ru ≥ 10.2%, Premion®
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
Dichlorotris(triphenylphosphine)ruthenium(II), Dichlorotris(triphenylphosphino)ruthenium (II)
Purity: 99.95%
CAS No.: 15529-49-4
Molecular Formula: RuCl2[P(C6H5)3]3
Formula Weight: 958.86
Physical Form: Crystalline powder
Sensitivity: Air and Moisture Sensitive
Appearance: Brown to black
Melting Point: 132-134°
Solubility: Very slightly soluble (with dissociation) in acetone, alcohol, chloroform, ethyl acetate, and toluene
MDL No.: MFCD00013077
Formule:
C₅₄H₄₅Cl₂P₃Ru Point de fusion: 132…134 °C Température de stockage: Température ambiante |
Numéro MDL:
MFCD00013077 Numéro CAS: 15529-49-4 EINECS: 239-569-7 |