Imprimer
Stoppers, rubber, Suba-Seal
Fournisseur: LEY RUBBER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TO/29/RED_U TO/45/CREAM TO/17/RED TO/45/RED TO/25/CREAM TO/37/REDU
217-0721EA 57.7 EUR
217-0721 LEYRTO/45/CREAM LEYRTO/17/RED LEYRTO/45/RED LEYRTO/25/CREAM LEYRTO/37/REDU
Stoppers, rubber, Suba-Seal
Bouchons Bouchons en caoutchouc
Turnover stoppers create a very effective double seal for narrow aperture containers.
  • No smell or taste
  • Will not yellow over time
  • Higher tear strength with less vulnerability to splitting or mechanical damage
External serrations create an excellent internal seal with the flexible turnover sleeve creating a secure seal on the container's outer neck. The head is easily pierced with a hypodermic needle.