Imprimer
Strips of 8 PCR tubes and cap strips
Fournisseur: RATIOLAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8610002 8610003 8610015
RATI8610002EA 119 EUR
RATI8610002 RATI8610003 RATI8610015
Strips of 8 PCR tubes and cap strips
Tubes Tubes PCR
Pure polypropylene.
  • Rapid and homogeneous heat transfer due to consistently ultrathin wall design
  • Perfect fit, compatible with all major brands of thermocyclers due to precise production
  • DNase-, RNase-, DNA and endotoxin-free
  • Variety of tight closures available to reduce sample evaporation
  • With attached domed cap strips or for use with detached 8-cap strips
  • Caps designed for tight sealing, easy to open and to close