Imprimer
Tubes PCR, 0,2 ml et 0,5 ml avec bouchon plat ou bombé
Fournisseur: RATIOLAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8610001 8610006 8610007
RATI8610001EA 55.3 EUR
RATI8610001 RATI8610006 RATI8610007
Tubes PCR, 0,2 ml et 0,5 ml avec bouchon plat ou bombé
Tubes Tubes PCR
Pure polypropylene.
  • Rapid and homogeneous heat transfer due to consistently ultrathin wall design
  • Perfect fit, compatible with all major brands of thermocyclers due to precise production
  • DNase-, RNase-, DNA and endotoxin-free
  • Variety of tight closures available to reduce sample evaporation
Caps designed for tight sealing, easy to open and to close.