Imprimer
Entonnoirs cytologiques jetables, CytoSep™, pour cytocentrifugeuse Shandon Cytospin® 4
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
M964-10FW M964-20FW M965-10FW M965-20FW M965-10FT M964-10FT M964-20FW1
720-1863EA 946 EUR
720-1863 720-1864 720-1865 720-1866 720-1948 720-1945 720-2947
Entonnoirs cytologiques jetables, CytoSep™, pour cytocentrifugeuse Shandon Cytospin® 4
Entonnoirs Chambres cytologiques
These one-way cytology funnels / cytofunnels are used for applying a thin cell layer to a microscope slide by means of centrifugation. The application area for cells has a diameter of about 6 mm (about 28 mm²). We offer both versions: For use with a reusable metal clip, as well as a version with integrated plastic base as a complete immobilising solution, e.g. In infectious samples.
  • For use without metal clips in the Shandon CytoSpin® centrifuge as well as in compatible rotors
  • Supplied plastic base (instead of metal clip) is a complete solution, for
En savoir plus