Imprimer
Standard sockets and cones, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
286150403 286151005 286151802 286152404 286155007 286156209 286159609 286160207 286160404 286160601 286161203 286161409 286161606 286161803 286162002 286514608 286515604 286517005 286520007 286536602 286543803 286559207 286563402 286564407 286565206 286565806 286566202 286525408 286550001 286561204 286561401 286561607 286153203 286157008
SCOT286150403EA 96.2 EUR
SCOT286150403 SCOT286151005 SCOT286151802 SCOT286152404 SCOT286155007 SCOT286156209 SCOT286159609 SCOT286160207 SCOT286160404 SCOT286160601 SCOT286161203 SCOT286161409 SCOT286161606 SCOT286161803 SCOT286162002 SCOT286514608 SCOT286515604 SCOT286517005 SCOT286520007 SCOT286536602 SCOT286543803 SCOT286559207 SCOT286563402 SCOT286564407 SCOT286565206 SCOT286565806 SCOT286566202 SCOT286525408 SCOT286550001 SCOT286561204 SCOT286561401 SCOT286561607 SCOT286153203 SCOT286157008
Standard sockets and cones, DURAN®
Adaptateurs Raccords à rodage conique
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes