Imprimer
Standard diamond ground joints, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
286155401 286156303 286518001 286521003 286541202 286546003 286532806 286558202 286154208 286158801
SCOT286155401EA 276 EUR
SCOT286155401 SCOT286156303 SCOT286518001 SCOT286521003 SCOT286541202 SCOT286546003 SCOT286532806 SCOT286558202 SCOT286154208 SCOT286158801
Standard diamond ground joints, DURAN®
Adaptateurs Raccords à rodage conique
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With Diamond ground.