Imprimer
Stirrer bearings, with vacuum jacket, KPG®
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
06302029 06302045
LENZ06302029EA 124 EUR
LENZ06302029 LENZ06302045
Stirrer bearings, with vacuum jacket, KPG®
Agitateurs Outils d'agitation
Stirrer bearings with NS cone and vacuum connections which made of precision calibrated glass tube.
  • Surface is grounded and polished
  • Interchangeable