Imprimer
Wash cloths, Tork
Fournisseur: Essity
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
742200 747400 747450
115-2403EA 167 EUR
115-2403 SCAA747400 SCAA747450
Wash cloths, Tork
Lingettes Lingettes et essuie-mains à usage unique Lingettes en papier
High quality wash cloths, ideal for washing, showering and general body care. They can be used with soap and water.
  • Highly absorbent and soft
  • Can be sterilized
  • Tork Premium: Extra soft for sensitive skin and extra wet-strong
Colour: White