Imprimer
Measuring cylinders tall form, without base, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
213931409 213931709 213933607 213934406 213935402 213930807 213932405
215-3625EA 84.1 EUR
215-3625 215-3626 215-3628 215-3629 215-3630 215-3624 215-3627
Measuring cylinders tall form, without base, DURAN®
Éprouvettes Éprouvettes graduées
DURAN®, borosilicate glass 3.3, clear.
  • Tall form
  • Flat bottom
  • With spout
  • Without base
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
  • Without foot