Imprimer
Stopcocks for separarting funnels, Gilson type, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
286428407 286428801 286429609 286429806 286430001
SCOT286428407EA 833 EUR
SCOT286428407 SCOT286428801 SCOT286429609 SCOT286429806 SCOT286430001
Stopcocks for separarting funnels, Gilson type, DURAN®
Stopcocks
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE key.
  • Very good chemical resistance
  • Very good temperature resistance
For glass manipulators.