Imprimer
Tubing connectors, X-shaped, LaboPlast®
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8708-0004 8708-0006 8708-0008 8708-0010 8708-0012 8708-0014 8717-0004 8717-0006 8717-0008 8717-0012 8717-0014 8717-0010
BURK8708-0004EA 10.8 EUR
BURK8708-0004 BURK8708-0006 BURK8708-0008 BURK8708-0010 BURK8708-0012 BURK8708-0014 229-0210 229-0213 229-0214 229-0216 229-0217 229-0215
Tubing connectors, X-shaped, LaboPlast®
Raccords et connecteurs
PP or PVDF, transparent.
  • PP, temperature resistant up to +135 °C max.
  • PVDF, temperature resistant up to +160 °C max., highly resistant to chemicals
  • Sterilisable
Cylindrical four-way hose connector to connect hoses. PVDF connectors with food industry approval, autoclavable and UV-resistant.