Imprimer
Tubing connectors, straight, universal, LaboPlast®
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8700-0417 8710-0417
BURK8700-0417EA 26.2 EUR
BURK8700-0417 229-0225
Tubing connectors, straight, universal, LaboPlast®
Raccords et connecteurs
PP or PVDF, translucent.
  • PP, temperature resistant up to +135 °C max.
  • PVDF, temperature resistant up to +160 °C max., highly resistant to chemicals
  • Sterilisable
Conical, straight hose connectors for universal use. PVDF connectors with food industry approval, autoclavable and UV-resistant.