Imprimer
Cell counter vials
Fournisseur: RATIOLAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5515040 5515010 5515041 5515051
RATI5515040EA 0 EUR
RATI5515040 RATI5515010 RATI5515041 RATI5515051
Cell counter vials
Fioles
Cell counter vials made of clear PS.
  • High-precision design for use with robotic analytical systems