Imprimer
Microtubes à centrifuger
Fournisseur: Ratiolab
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ratiolab®
211-2065EA 19.1 EUR
211-2065 RATI5615001 RATI5615002 RATI5615101 RATI5615003 RATI5615004 RATI5615400 RATI5615020 RATI5615000
Microtubes à centrifuger
Tubes Microtubes à centrifuger
PP, translucent.
  • Autoclavable with opened cap at 121 °C, 20 min
  • Natural colour or different colours for comfortable identification
  • Tight sealing cap
  • Safe and easy to be opened and closed
With or without flat cap.