Imprimer
Centrifuge tubes, conical bottom
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
T420-7 T420-6 T420-1 T420-3 T420-4 T420-5
SIMPT420-7EA 197 EUR
SIMPT420-7 SIMPT420-6 SIMPT420-1 SIMPT420-3 SIMPT420-4 SIMPT420-5
Centrifuge tubes, conical bottom
Tubes Tubes à centrifuger
Tubes made of either polystyrene, optically clear or polypropylene, translucent.
  • Molded graduations from 2,5 to 50 ml
  • Polypropylene tubes can be autoclaved
  • Sterile or non sterile available
These centrifuge tubes are also useful for collecting and transporting biological specimens. Leakproof characteristics are ensured by a flat top plastic screw cap with an inner sealing lip.
Informations de commande: Tubes are available in bags or in polypropylene racks for better protection during transport, storage and for convenient laboratory use. Racks can hold up to 25 tubes.
Informations de livraison: Tubes are supplied sterile with green caps or non sterile with yellow caps.