Imprimer
Tubing connectors, complete with screw cap, straight, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
286230005 286230202 286230408
SCOT286230005EA 11.3 EUR
SCOT286230005 SCOT286230202 SCOT286230408
Tubing connectors, complete with screw cap, straight, DURAN®
Raccords et connecteurs
Screw cap PTB, red, with white PTFE straight hose connection.
  • Very good chemical resistance
  • Very good temperature resistance
With silicone seal (VMQ).