Imprimer
Double bossheads
Fournisseur: BOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5300 5305 5310
241-3028EA 68.8 EUR
241-3028 451-9111 BOCH5310
Double bossheads
Pinces Noix de serrage
Double bossheads made of stainless steel 18/10.
  • For rods up to 16,5 mm
  • Angle: 90°
  • Available with heart screw M6/M8, socket screw M6/M8 or safety screw type 'heart' M8
Certifications: DIN 12895