Imprimer
Conteneurs de transport sécurisés
Fournisseur: BOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8300SS 8301SS 18301SS 18300SS
BOCH8300SSEA 425 EUR
BOCH8300SS BOCH8301SS BOCH18301SS BOCH18300SS
Conteneurs de transport sécurisés
Portoirs Portoirs pour flacons
Safety transport container, made of stainless steel, for laboratory bottles.
  • For bottles 50 to 1000 ml
  • With hinge, lock and silicone seal
  • Good chemical resistance