Imprimer
Portoirs pour microtubes, OneHand™, polymère acétal
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
T360-50B T360-50G T360-50O T360-50Y T360-50L
SIMPT360-50BEA 182 EUR
SIMPT360-50B SIMPT360-50G SIMPT360-50O SIMPT360-50Y SIMPT360-50L
Portoirs pour microtubes, OneHand™, polymère acétal
Portoirs Tube Racks
These microtube racks are made of highly resistant acetal as it will not shatter or stain in contact with most laboratory chemicals.
  • Most compacted, lightweight and stackable
  • Ideal for incubators, refrigerators, freezers under lab hoods and on bench tops
  • Interlocking feet allow safe stacking
  • Convenient handling on each side of the rack will ensure a safe grip when carrying it around
The OneHand™ microtube racks is made of two tiers to facilitate the insertion and stability of microtubes. They are available
in five different colors that are individually wrapped.