Imprimer
Microtubes à centrifuger, SnapTwist™ Micrewtube®
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
T342-5T T342-4T T342-6T T342-7T
SIMPT342-5TEA 85.1 EUR
SIMPT342-5T SIMPT342-4T SIMPT342-6T SIMPT342-7T
Microtubes à centrifuger, SnapTwist™ Micrewtube®
Tubes Microtubes à centrifuger
PP, translucent, with PE screw cap.
  • Without graduations
  • Caps cannot be autoclaved since they are made of high density polyethylene
  • Conical bottom tubes can be centrifuged up to 20000×g
  • Skirted tubes can be centrifuged up to 17000×g
  • Temperature range: -90 to +100 °C
  • Certified RNase-, DNase-, Pyrogen- and DNA-free
The SnapTwist™ Micrewtube® provides all the advantages of a modern microcentrifuge tube with screw cap but the closure is a true time saver. The SnapTwist™ Micrewtube® has a multitude of applications around your lab. It is ideal for freezer storage, boiling application, centrifugation etc, and will fit most standard microcentrifuge rotors.
The tubes can be securely sealed by simply capping the closures on. Removal of caps requires an easy 1/4 turn (twist). The ease with which these caps can be manipulated eliminates the danger of spillage associated with other push-on/pull-off caps. The deep internal lip of the cap fits snugly against the interior wall of the tube preventing the contents from coming in contact with the threads, thus reducing the chances of sample contamination. The quality of the sealing system is such that it is not necessary to tighten the closure with pressure to achieve a leakproof seal.