Imprimer
Flacons d'échantillons dans Lab File®, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
224881 224882 224884 224886 224888 224981 224984 225288 225081 224982 225012 225014 224885 225289 225290
548-2135EA 545 EUR
548-2135 548-2136 548-2137 548-2423 548-2139 218-2211 218-2213 548-2187 218-0004 218-2212 548-2130 548-2131 548-2138 548-2424 548-2425
Flacons d'échantillons dans Lab File®, WHEATON®
Vials
Clear vials made from low extractable borosilicate glass, amber vials made from borosilicate glass.
  • Sample vials with caps attached to vial help maintain cleanliness
  • Lab File® with partitioned trays provides an easy way to inventory samples or to store empty vials
  • Clear vials conform to ASTM E438 Type I, Class A and USP Type I requirements
  • Amber vials conform to USP Type I requirements for light transmission to protect light-sensitive products
  • 14B rubber lined black phenolic screw cap for aqueous samples