Imprimer
Flow through coolers, with natural refrigerant DC®
Fournisseur: HUBER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3000.0003.00 3001.0003.00
HUBE3000.0003.00EA 3360 EUR
HUBE3000.0003.00 HUBE3001.0003.00
Flow through coolers, with natural refrigerant DC®
Thermostats à circulation Systèmes à circulation fermée Circulateurs de refroidissement
Flow-through chillers are designed for simple, low demand cooling applications. They are commonly used in combination with heating circulators to remove heat in order to cool a process back to room temperature.
  • Ideal for counter-cooling or as a replacement for cooling water
  • Compact, space-saving construction
  • Robust housing with carrying grips
  • Environmentally friendly with natural refrigerant
  • Natural refrigerant
Informations de commande: Accessories included. There are also some optional accessories.
Attention: Cooling capacity at +15; 0; −20 °C.