Imprimer
Screwthread tubes, attachable, DIN thread, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
248350103 248380209 248360207 248370105 248390107
SCOT248350103EA 52.6 EUR
SCOT248350103 SCOT248380209 SCOT248360207 SCOT248370105 SCOT248390107
Screwthread tubes, attachable, DIN thread, DURAN®
Adaptateurs Adaptateurs pour tubes filetés
Made of DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.
DIN 12216