Imprimer
Sponge, VertiKlean®
Fournisseur: CONTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VKSPONGE-1 VKST0003
115-0179EA 225 EUR
115-0179 115-3203
Sponge, VertiKlean®
Wipes Sponges
The VertiKlean Sponge is a disposable laboratory sponge. It consists of Polynit wiping material that has been laminated to a clean polyester foam. The sponge is laser cut to completely seal the edges. It is designed for general cleaning and wiping but can also be used in laboratories.
  • High sorbency polyester foam with the abrasion resistance of Polynit fabric
  • Autoclavable