Imprimer
Funnels, disposable, Microsart® 100/250
Fournisseur: Sartorius
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
16A07-10-N 16A07-25-N
SART16A07-10-NEA 173 EUR
SART16A07-10-N 514-0190
Funnels, disposable, Microsart® 100/250
Funnels Membrane Filter Funnels
PP housing.
  • Reliable results-use a new, sterile funnel for each test for certain avoidances of cross contamination
  • Time saving-kust change the funnel, rather than spending time sanitizing it
  • Simpler handling-no more holding of hot funnels. And you can see when filtration has been completed, particulary useful when using manifolds in routine testing
Microsart® funnels are sterile 100 ml and 250 ml respectively plastic funnels. They allow quick performance of the filtrations required in the routine testing of water, food and beverages, pharmaceutical and cosmetic products.