Imprimer
Microtubes à centrifuger, 2,0 ml, Cliklok™
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
T330-72G T330-72N T330-72AN
SIMPT330-72GEA 0 EUR
SIMPT330-72G SIMPT330-72N SIMPT330-72AN
Microtubes à centrifuger, 2,0 ml, Cliklok™
Tubes Microtubes à centrifuger
Made of polypropylene, translucent with snap cap.
  • Graduated at 0,5, 1,0 and 1,5 ml
  • Certified RNase, DNase, pyrogen and DNA-free
  • Pierceable lid
  • Withstand centrifugation up to 15 000×g
  • Autoclavable
These 2 ml microcentrifuge tubes offer a special design to produce a more secure closure. This will help to prevent tubes from opening during centrifugation, boiling, storing, freezing and shipping. Etched surface on side of tube for sample identification. Large etched graduations make volumes easy to read. Improved polypropylene transparency for easy viewing of samples.