Imprimer
Plaques pour PCR 96 puits, opaques, à jupe, Amplate™
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
T324-96SKK T324-96SKW
SIMPT324-96SKKEA 384 EUR
SIMPT324-96SKK SIMPT324-96SKW
Plaques pour PCR 96 puits, opaques, à jupe, Amplate™
Microplaques Plaques PCR
PP. These opaque 96-well PCR plates are for chemiluminescent and fluorescent procedures. Thin-walled and designed for rapid thermal transfer, these plates are precision-moulded to ensure well-to-well and plate-to-plate uniformity.
  • Opaque to ensure low level of background fluorescence
  • Alphanumeric grid helps sample identification
  • Corner at H1 of the plate is cut away to facilitate orientation
  • Easy to seal since no cylindrical walls extend above the plate
  • Area for bar coding, labelling or writing
Can be handled by robotic handling equipment and is ideal with automated pipetting systems.
Informations de livraison: Packed in tamperproof resealable bags of 10 plates.