Imprimer
Spherical ground joints, cup, DURAN®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
286320208 286321007 286322209 286322603 286323805 286324201 286326605 286327001 286327207 286328203 286329002 286325403
SCOT286320208EA 943 EUR
SCOT286320208 SCOT286321007 SCOT286322209 SCOT286322603 SCOT286323805 SCOT286324201 SCOT286326605 SCOT286327001 SCOT286327207 SCOT286328203 SCOT286329002 SCOT286325403
Spherical ground joints, cup, DURAN®
Adaptateurs Adaptateurs rodés sphériques
DURAN®, borosilicate glass 3.3, clear.
  • Inches size
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Through use of flexible spherical joint connections, apparatus can be assembled free from stresses.
*non-DIN size
Certifications: DIN 12244, DIN 12264