Imprimer
Safety gas burners, with needle valve
Fournisseur: BOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7410S 7400S
451-1401EA 262 EUR
451-1401 BOCH7400S
Safety gas burners, with needle valve
Appareils chauffants Brûleurs à gaz
Safety gas burner with needle valve and anti-slip rubber coating under the base.
  • Thermo safety element
  • Pilot flame
  • Air regulation
  • Max. 1300 °C