Imprimer
Burners, Teclu
Fournisseur: BOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7150 7160
BOCH7150EA 204 EUR
BOCH7150 BOCH7160
Burners, Teclu
Appareils chauffants Brûleurs à gaz
These burners reach a maximum temperature of 1300 °C and have an anti-slip rubber coating under the base.
  • With air regulation
  • With stopcock
  • Pilot flame