Imprimer
Samplers, pigment lance
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5369-0300 5369-0900 5369-0610 5369-0910 5369-0600 5369-0310
231-0480EA 295 EUR
231-0480 231-0484 231-0483 231-0485 231-0482 231-0481
Samplers, pigment lance
Samplers Solid Samplers
Pigment lance to sample pigments, pastes and agglomerating bulk goods.
  • Half-open flute, 50%, for highly sticky products
  • Quarter-open flute, 25%, for less sticky products
  • Stainless steel V4A (1.4404)