Imprimer
Échantillonneurs, lance vanne
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5359-1600 5359-1800
300-0076EA 1980 EUR
300-0076 300-0077
Échantillonneurs, lance vanne
Échantillonneurs Préleveurs de solides
Samplers for viscous, paste-like and semi-liquid substances.
  • Optimised opening mechanism (can be completely disassembled)
  • Easy to use and to clean
  • Different layers remain intact particularly with paste-like substances
Designed for use in pharmaceutical applications.
Certifications: DIN 51750.3 and 53242.3