Imprimer
Plaques à puits profonds, 96 puits, Nunc™
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
T110-3 T110-2
SIMPT110-3EA 260 EUR
SIMPT110-3 SIMPT110-2
Plaques à puits profonds, 96 puits, Nunc™
Microplaques Microplaques deepwell
All BioBlock™ Deepwell Plates are ideal for high volume, high throughput applications. The high-performance plates are chemical-resistant and designed for automated robotic handling.
  • Footprint conforms to SBS/ANSI standards
  • Great chemical stability
  • Free of RNase, DNase, Pyrogen, and DNA
  • Maximum sustainable centrifugal force up to 4000 g
  • The height difference from center to angle is less than 0,4 mm