Imprimer
Tapis de scellage pour plaques à puits profonds Bioblock™
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
T110-25 T110-26
SIMPT110-25EA 158 EUR
SIMPT110-25 SIMPT110-26
Tapis de scellage pour plaques à puits profonds Bioblock™
Scellés Microplaque Sceaux
Designed to fit the Bioblock™ family, these mat covers are made of a specially formulated plastic ensuring great flexibility. They allow for maximum sample volume in each well. They are resistant to DMSO and biological buffers.
Polyoelefin and elastomer mats can be used within a temperature range of −80 to +80 °C.