Imprimer
Plaque 384 puits à puits carrés profonds, Bioblock™
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
T110-200 T110-7
SIMPT110-200EA 292 EUR
SIMPT110-200 SIMPT110-7
Plaque 384 puits à puits carrés profonds, Bioblock™
Microplaques Microplaques deepwell
PP. This 384-well plate is ideal for compound storage and handling of biological samples.
  • Round well bottom facilitates aspiration
  • Suitable to be used with DMSO and biological buffers
Informations de livraison: Packed in a bag of 4 plates.