Imprimer
Plasma separator tubes, PST™ II, BD Vacutainer®
Fournisseur: BD Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
367376 367374 367378
BDAM367376EA 210 EUR
BDAM367376 BDAM367374 BDAM367378
Plasma separator tubes, PST™ II, BD Vacutainer®
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons de sang
Plasma tubes with separating gel for clinical chemistry are available with spray-dried lithium heparin additives.
  • It act as stable barrier between plasma and cells providing analyte stability and better sample quality
  • Correct mixing of the tubes immediately after the blood sample collection is important to avoid micro clotting
  • Plasma separation tubes should be gently inverted 180° and back 8 to 10 times
During the centrifugation of these tubes, an inert gel separates the plasma and the cells preventing the contamination of the plasma from the separated cellular components.
Informations de commande: Study documentation in relation to BD Vacutainer® PST™ II tubes are supplied on request.