Imprimer
Pelles pharmaceutiques
Fournisseur: BOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8810 8811 8812 8813 8809
BOCH8810EA 154 EUR
BOCH8810 231-0205 BOCH8812 BOCH8813 BOCH8809
Pelles pharmaceutiques
Pelles
These chemical scoops made from stainless steel 18/10 are ideal for use in pharmaceutical and food industries.
  • With welded handle
  • Food contact approved