Imprimer
Evaporating dishes with spout
Fournisseur: BOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8023 8020 8021 8022 8024
BOCH8023EA 94.6 EUR
BOCH8023 BOCH8020 BOCH8021 BOCH8022 BOCH8024
Evaporating dishes with spout
Plateaux/Récipients/Pots Capsules d'évaporation
Evaporating dish, made of nickel, with spout.
  • Corrosion resistant
  • Very good thermal resistance
  • Smooth surfaces for easy cleaning
Multipurpose tray ideal for a wide variety of uses.