Imprimer
Evaporating dishes, high shape
Fournisseur: BOCHEM
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8560 8565 8561 8563 8562 8564
BOCH8560EA 39.4 EUR
BOCH8560 BOCH8565 BOCH8561 BOCH8563 BOCH8562 BOCH8564
Evaporating dishes, high shape
Plateaux/Récipients/Pots Capsules d'évaporation
Evaporating dish, stainless steel, with contact bottom for heating plates and magnetic stirrers.
  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable