Imprimer
Soil samplers, Mole
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5350-1045 5350-1005 5350-1015 5350-1020 5350-1025 5350-1030 5350-1035 5350-1040
BURK5350-1045EA 560 EUR
BURK5350-1045 BURK5350-1005 BURK5350-1015 BURK5350-1020 BURK5350-1025 BURK5350-1030 BURK5350-1035 BURK5350-1040
Soil samplers, Mole
Samplers Solid Samplers
Very robust model with specially hardened forged drill bits. For determining soil properties, moisture and structure. The shape of the Mole‘s drill bits ensures minimum friction when drilling and removing the Mole from the ground.The Mole has different components. Handle with 75 cm rod, extension rod 100 cm if necessary and one of seven available drill bits, each of which is designed for a particular soil type. The components can be combined without any problems. Extension rods can extend the working depth of the Mole down to 5 m. Please order drill bits seperately.
  • Physical efforts are reduced to a minimum