Imprimer
Media bottles, graduated, Lab 45™, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
219927 219931 219929 219930 219917 219919 219920 219921
WHEA219927EA 203 EUR
WHEA219927 WHEA219931 WHEA219929 WHEA219930 WHEA219917 WHEA219919 WHEA219920 WHEA219921
Media bottles, graduated, Lab 45™, WHEATON®
Flacons Flacons pour milieux de culture
Borosilicate glass, USP type I.
  • Graduated with writing patch
  • Bottles, caps and pour rings are autoclavable
  • Cap has a built-in wedge shaped sealing ring, eliminating the need for a liner
Wide mouth media storage bottles are designed for heavy duty storage as well as mixing and sampling.