Imprimer
Media bottles, graduated, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
219875 219845 219855 219865
WHEA219875EA 1050 EUR
WHEA219875 WHEA219845 WHEA219855 WHEA219865
Media bottles, graduated, WHEATON®
Flacons Flacons pour milieux de culture
Amber borosilicate glass bottle with graduations and marking patch.
  • Graduations and marking patch assists in measuring volume and identifying content
  • Special pour lip facilitates pouring
  • Packaged in partitioned carton to help prevent scratch marks on bottles
Informations de commande: Black phenolic screw cap can be purchased separately