Imprimer
Clamps, with sleeve
Fournisseur: RSG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
451.625 451.640 451.660
RSGA451.625EA 56.3 EUR
RSGA451.625 RSGA451.640 RSGA451.660
Clamps, with sleeve
Pinces Pinces à usage général
These clamps with sleeve are made of electrolytically polished stainless steel.
  • Stable clamping screws
  • Jaws fitted with cork
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable