Imprimer
Media bottles, safety-coated, without screw cap, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
219457 219455
WHEA219457EA 686 EUR
WHEA219457 WHEA219455
Media bottles, safety-coated, without screw cap, WHEATON®
Flacons Flacons pour milieux de culture
USP Type I clear borosilicate glass, Plastisol coated.
  • Provided with graduations and writing patch
  • 33-430 screw cap can be purchased separately
  • Autoclavable