Imprimer
Pelles, EasyScoop
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5356-1500 5356-2000
BURK5356-1500EA 53.9 EUR
BURK5356-1500 BURK5356-2000
Pelles, EasyScoop
Pelles
Sampling scoops made of light aluminium, bright-anodised.
  • Ergonomically designed: With centrally arranged inner grip
  • Considerably shorter than conventional scoops: Easier to take samples from narrow containers
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable